Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOCOS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589769 Supplier Invitrogen™ Supplier No. PA589769
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIKAKGMEPI EVPLEENSER TQIRQSRVCA DRVSTYDCGE KISSWLSTFF GRPCHLIKQS SNSQRNAKKK HGKDQLPGTM ATLSLVNEAQ YLLINTSSIL ELHRQLNTSD ENGKEELFSL KDLSLRFRAN IIINGKRAFE EEKWDEISIG SLRFQVLGPC HRCQMICIDQ QTGQRNQHVF QKLSESRETK VNFGMYLMHA SLDLSSPCFL SVGSQVLPVL KENVEGHDLP ASEKHQDVTS; Positive Samples: HepG2.

MOCOS sulfurates the molybdenum cofactor of xanthine dehydrogenase (XDH), which is required for their enzymatic activities (Ichida et al., 2001).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOCOS
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MOCOS
Gene Accession No. Q96EN8
Gene Alias 1110018O12Rik; HMCS; MCS; moCo sulfurase; MOCOS; Molybdenum cofactor sulfurase; molybdenum cofactor sulfurtransferase; MOS
Gene Symbols MOCOS
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 649-888 of human MOCOS (NP_0604172).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55034
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.