Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOCOS Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569105
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9105 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9105 Supplier Bethyl Laboratories, Inc Supplier No. A304949AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

MOCOS sulfurates the molybdenum cofactor of xanthine dehydrogenase (XDH; MIM 607633) and aldehyde oxidase (AOX1; MIM 602841), which is required for their enzymatic activities (Ichida et al., 2001 [PubMed 11302742]).

Specifications

Antigen MOCOS
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene MOCOS
Gene Accession No. Q96EN8
Gene Alias HMCS, MCS, MOS, moCo sulfurase
Gene Symbols MOCOS
Host Species Rabbit
Immunogen Between 838 and 888
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55034
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.