Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOCS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567387 Supplier Invitrogen™ Supplier No. PA567387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSMSEHFCGTC NRLRITADGN LKVCLFGNSE VSLRDHLRAG ASEQELLRII GAAVGRKKRQ HAGMFSISQM KN Highest antigen sequence identity to the following orthologs - mouse 96%, rat 95%.

MONDOA forms heterodimers with MLX (MIM 602976) that can bind to and activate transcription from CACGTG E boxes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOCS1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MOCS1
Gene Accession No. Q9NZB8
Gene Alias 3110045D15Rik; AI043170; AW536397; bHLHe36; Cell migration-inducing gene 11 protein; Cyclic pyranopterin monophosphate synthase; GTP 3',8-cyclase; MIG11; migration-inducing gene 11 protein; MIR; Mlx interactor; MOCOD; MOCODA; MOCS1; MOCS1A enzyme; molybdenum cofactor biosynthesis protein 1; Molybdenum cofactor biosynthesis protein A; Molybdenum cofactor biosynthesis protein C; molybdenum cofactor synthesis 1; molybdenum cofactor synthesis-step 1 protein A-B; molybdenum cofactor synthesis-step 1 proteins A and B; MONDOA
Gene Symbols MOCS1
Host Species Rabbit
Immunogen Recombinant Human MOCS1. Recombinant protein control fragment (Product #RP-108048).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4337
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.