Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MOCS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113418
Description
MOCS1 Polyclonal Antibody for Western Blot, ELISA
MONDOA forms heterodimers with MLX (MIM 602976) that can bind to and activate transcription from CACGTG E boxes.
Specifications
MOCS1 | |
Polyclonal | |
Unconjugated | |
MOCS1 | |
3110045D15Rik; AI043170; AW536397; bHLHe36; Cell migration-inducing gene 11 protein; Cyclic pyranopterin monophosphate synthase; GTP 3',8-cyclase; MIG11; migration-inducing gene 11 protein; MIR; Mlx interactor; MOCOD; MOCODA; MOCS1; MOCS1A enzyme; molybdenum cofactor biosynthesis protein 1; Molybdenum cofactor biosynthesis protein A; Molybdenum cofactor biosynthesis protein C; molybdenum cofactor synthesis 1; molybdenum cofactor synthesis-step 1 protein A-B; molybdenum cofactor synthesis-step 1 proteins A and B; MONDOA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4337, 56738 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5RKZ7, Q9NZB8 | |
MOCS1 | |
Recombinant Human Molybdenum cofactor biosynthesis protein 1 protein (90-360AA). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction