Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOCS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558054 Supplier Invitrogen™ Supplier No. PA558054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTRNNFEGKK VISLEYEAYL PMAENEVRKI CSDIRQKWPV KHIAVFHRLG LVPVSEASII IAVSSAHRAA SLEAVSYAID TLKAKVP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

Eukaryotic molybdoenzymes use a unique molybdenum cofactor (MoCo) consisting of a pterin, termed molybdopterin, and the catalytically active metal molybdenum. MoCo is synthesized from precursor Z by the heterodimeric enzyme molybdopterin synthase. The large and small subunits of molybdopterin synthase are both encoded from this gene by overlapping open reading frames. The proteins were initially thought to be encoded from a bicistronic transcript. They are now thought to be encoded from monocistronic transcripts. Alternatively spliced transcripts have been found for this locus that encode the large and small subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOCS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MOCS2
Gene Accession No. O96007, O96033
Gene Alias AI415403; HAMAP-Rule:MF_03052}; MCBPE; MOCO1; MOCO1-A; MOCO1-B; MOCODB; Mocs2; mocs2 {ECO:0000255; MOCS2A; MOCS2B; MOCS2B {ECO:0000255; molybdenum cofactor; molybdenum cofactor biosynthesis protein E; molybdenum cofactor synthesis 2; molybdenum cofactor synthesis protein 2 large subunit; molybdenum cofactor synthesis protein 2 large subunit {ECO:0000255; Molybdenum cofactor synthesis protein 2 small subunit; Molybdenum cofactor synthesis protein 2A; Molybdenum cofactor synthesis protein 2B; molybdenum cofactor synthesis protein 2B {ECO:0000255; molybdopterin synthase catalytic subunit; molybdopterin synthase catalytic subunit {ECO:0000255; molybdopterin synthase catalytic subunit; molybdopterin synthase sulfur carrier subunit; molybdopterin synthase small and large subunit; molybdopterin synthase sulfur carrier subunit; Molybdopterin-synthase large subunit; Molybdopterin-synthase small subunit; MPT synthase large subunit; MPTS; Sulfur carrier protein MOCS2A
Gene Symbols MOCS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MOCS2. Recombinant protein control fragment (Product #RP-102780).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4338
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.