Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOCS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109968 Supplier Invitrogen™ Supplier No. PA5109968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSLEISSSC FSLETKLPLS PPLVEDSAFE PSRKDMDEVE EKSKDVINFT AEKLSVDEVS QLVISPLCGA ISLFVGTTRN NFEGKKVISL EYEAYLPMAE NEVRKICSDI RQKWPVKHIA VFHRLGLVPV SEASIIIAVS SAHRAASLEA VSYAIDTLKA KVPIWKKEIY EESSTWKGNK ECFWASNS.

Eukaryotic molybdoenzymes use a unique molybdenum cofactor (MoCo) consisting of a pterin, termed molybdopterin, and the catalytically active metal molybdenum. MoCo is synthesized from precursor Z by the heterodimeric enzyme molybdopterin synthase. The large and small subunits of molybdopterin synthase are both encoded from this gene by overlapping open reading frames. The proteins were initially thought to be encoded from a bicistronic transcript. They are now thought to be encoded from monocistronic transcripts. Alternatively spliced transcripts have been found for this locus that encode the large and small subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOCS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MOCS2
Gene Accession No. Q9Z224
Gene Alias AI415403; HAMAP-Rule:MF_03052}; MCBPE; MOCO1; MOCO1-A; MOCO1-B; MOCODB; Mocs2; mocs2 {ECO:0000255; MOCS2A; MOCS2B; MOCS2B {ECO:0000255; molybdenum cofactor; molybdenum cofactor biosynthesis protein E; molybdenum cofactor synthesis 2; molybdenum cofactor synthesis protein 2 large subunit; molybdenum cofactor synthesis protein 2 large subunit {ECO:0000255; Molybdenum cofactor synthesis protein 2 small subunit; Molybdenum cofactor synthesis protein 2A; Molybdenum cofactor synthesis protein 2B; molybdenum cofactor synthesis protein 2B {ECO:0000255; molybdopterin synthase catalytic subunit; molybdopterin synthase catalytic subunit {ECO:0000255; molybdopterin synthase catalytic subunit; molybdopterin synthase sulfur carrier subunit; molybdopterin synthase small and large subunit; molybdopterin synthase sulfur carrier subunit; Molybdopterin-synthase large subunit; Molybdopterin-synthase small subunit; MPT synthase large subunit; MPTS; Sulfur carrier protein MOCS2A
Gene Symbols MOCS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-188 of human MOCS2 (NP_004522.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17434
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.