Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOCS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597977 Supplier Invitrogen™ Supplier No. PA597977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MOCS3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Molybdenum cofactor (MoCo) is necessary for the function of all molybdoenzymes. One of the enzymes required for the biosynthesis of MoCo is molybdopterin synthase (MPT synthase). The protein encoded by this gene adenylates and activates MPT synthase. This gene contains no introns. A pseudogene of this gene is present on chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOCS3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MOCS3
Gene Accession No. O95396
Gene Alias 1700020H17Rik; Adenylyltransferase and sulfurtransferase MOCS3; Adenylyltransferase MOCS3; dJ914P20.3; MOCS3; molybdenum cofactor synthesis 3; molybdenum cofactor synthesis protein 3; Molybdopterin synthase sulfurylase; Molybdopterin-synthase adenylyltransferase; Molybdopterin-synthase sulfurtransferase; MPT synthase sulfurylase; Sulfur carrier protein MOCS2A adenylyltransferase; Sulfur carrier protein MOCS2A sulfurtransferase; Sulfurtransferase MOCS3; Uba4; UBA4, ubiquitin-activating enzyme E1 homolog; ubiquitin-like modifier activating enzyme 4
Gene Symbols MOCS3
Host Species Rabbit
Immunogen Recombinant Human Adenylyltransferase and sulfurtransferase MOCS3 protein (271-460AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27304
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.