Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Model Human Skull - Numbered 3 Parts PROMO
SCI_ED

Catalog No. S15581W25
Click to view available options
:
3 Parts

Human skull model replica.

  • Each part on the skull is numbered
  • Jaw is spring loaded
  • Removable Skull cap
  • Skull is made of durable plastic

Specifications

Product Type Human Skull Model
Dimensions (L x W x H) 8.0 x 6.0 x 8.0 in.
Includes laminated sheet with each numbers name for identification
Material Durable Plastic
Weight (English) 2.2 lb.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.