Description
Includes
Female/male connection, regulating valve with gauge to measure cylinder pressure, sampling line, calibration test tubing, sensor adapter (where applicable), and hard carrying case with foam inserts for accommodating two calibration gas cylinders
Kits available with 0.25L/min. or 1.5L/min. regulators.
Order Info
Cylinders are sold separately. (Measure approximately 14.25L x 3.5 in. diameter.)
Certifications
All calibration accessories are certified and tested for accuracy.
Specifications
Specifications
| Type | Calibration Kit |
| Flow Rate | 0.25 L/min. |