Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Simple Animal Cell Model
SDPSCI_ED

Catalog No. S05720 Shop All EDVOTEK Products
Click to view available options
Grade:
4 to 12
This item is not returnable. View return policy
This item is not returnable. View return policy

The Origami Organelles Simple Animal Cell Model provides a fun introduction to animal cells for students of any age.

  • Your students will enjoy learning about cells with this colorful model. The model covers all the main parts of cells (cell membrane, nucleus, chromosomes, mitochondria). For more advanced students, you can extend the model with optional parts (rough ER, smooth ER, and Golgi).
  • Students make their cell model using the colorful parts working alone or in small groups. They can make a simple version or use the optional parts to show more features. It can be printed in color, or in mono on colored or white paper. Includes a full-color Powerpoint class presentation.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions:
17 cm x 14 cm (7 in. x 5 in.)

Download includes:
Instructions, model templates and PowerPoint file

All you need:
Paper, scissors, and tape

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology - Cells
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.