Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Device Modified Auramine O
SDP

Catalog No. p-4644463
Click to view available options
Quantity:
250mL
1 x 4L

Fluorescent acid fast stain in two minutes

  • Permanganate step is omitted, only two step staining technique for acid fast organisms saves lot of time
  • For all non-acid fast organisms and debris, new counterstain quenches fluorescence
  • Yields cleaner field of view

Specifications

Type Modified Auramine O Stain
Quantity 1 x 4L
Includes Stain and decolorizer or quencher
Format Bottle
For Use With (Application) Rapid and economical staining technique for acid fast organisms
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.