Learn More
StatLab™ Modified EA-50 Cytology Stain
Click to view available options
Quantity:
4 x 1 gal.
4 x 16 oz. Bottle
Description
- Contains twice as much Eosin-Y than EA-50 and no Bismarck Brown
- An alcohol-based counterstain used in the cytology laboratory for routine cytoplasmic staining
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | Cytology Stain |
| Quantity | 4 x 1 gal. |