Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Modular Serological Pipette Holder, 1 Pocket Unit
SDP

The Modular Serological Pipette Holder, 1 Pocket Unit is a supplementary unit for Modular Serological Pipette Holder (one column version).

$85.90 - $117.70

Specifications

Product Type Pipet Box Holder
Positions 1 Pocket
View More Specs

Products 3
Catalog Number Mfr. No. Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Price Quantity & Availability  
22-000-615
SDP
View Documents
Poltex
SRPIPSTAK125
2.5 in.
Each for $85.90
Only null left
Add to Cart
 
22-000-616
SDP
View Documents
Poltex
SRPIPSTAK135
3.5 in.
Each for $101.80
Only null left
Add to Cart
 
22-000-617
SDP
View Documents
Poltex
SRPIPSTAK145
4.5 in.
Each for $117.70
Only null left
Add to Cart
 
Description

Description

  • Choose from 2.5, 3.5, or 4.5 in. tall.
  • Attaches to themselves and/or Modular Serological Pipette Holder Starter Set (SRPIPSTAK1START).
  • See image for dimensions.
  • Comes with mounting hardware.
Specifications

Specifications

Pipet Box Holder
1 Pocket
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.