Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Moesin Antibody (rMSN/492), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308923MFV450
Description
Moesin Monoclonal antibody specifically detects Moesin in Human, Rat samples. It is validated for Flow CytometrySpecifications
Moesin | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Flow Cytometry | |
rMSN/492 | |
Flow Cytometry | |
Membrane-organizing extension spike protein, moesin | |
Recombinant full-length human Moesin protein (Uniprot: P26038) | |
0.1 mL | |
Cell Biology, Cellular Markers, Stem Cell Markers | |
4478 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction