Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Moesin Mouse anti-Human, Rat, Clone: MSN/491, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Moesin |
---|---|
Clone | MSN/491 |
Dilution | Western Blot 0.5-1ug/ml, Simple Western 1.0 ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, SDS-Page |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB3287520UG
![]() |
Novus Biologicals
NBP23287520UG |
20 μg |
Each for $268.00
|
|
|||||
NB32875200U
![]() |
Novus Biologicals
NBP232875200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Moesin Monoclonal specifically detects Moesin in Human, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Moesin | |
Western Blot 0.5-1ug/ml, Simple Western 1.0 ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
P26038 | |
4478 | |
Recombinant human Moesin protein | |
Primary | |
Store at 4C. |
MSN/491 | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cytoskeleton Markers, Stem Cell Markers | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
Membrane-organizing extension spike protein, moesin | |
MSN | |
IgG1 κ | |
Protein A purified | |
Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title