Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Moesin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120797 Supplier Invitrogen™ Supplier No. PA5120797
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, OVCAR3, HT-29, SW620, Mouse spleen, Mouse lung, Mouse kidney. The target is usually found in the following locations: Apical cell membrane, Cell membrane, Cell projection, Cytoplasm, Cytoplasmic side, Peripheral membrane protein, cytoskeleton, microvillus membrane. Immunogen sequence: MPKTISVRVT TMDAELEFAI QPNTTGKQLF DQVVKTIGLR EVWFFGLQYQ DTKGFSTWLK LNKKVTAQDV RKESPLLFKF RAKFYPEDVS EELIQDITQR LFFLQVKEGI LNDDIYCPPE TAVLLASYAV QSKYGDFNKE VHKSGYLAGD KLLPQRVLEQ HKLNKDQWEE RIQVWHEEHR GMLREDAVLE YLKIAQDLEM YGVNYFSIKN KKGSELWLGV DALGLNIYEQ NDRLTPKIGF PWSEIRNISF NDKKFVIKPI DKKAPDFVFY APRLRINKRI LALCMGNHEL YMRRRKPDTI.

Moesin, a member of the talin-4. 1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Moesin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MSN
Gene Accession No. P26038, P26041
Gene Alias C78546; epididymis luminal protein 70; HEL70; Membrane-organizing extension spike protein; MGC137877 protein; moesin; MSN
Gene Symbols MSN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human Moesin (NP_002435.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17698, 4478
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.