Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Moesin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582424 Supplier Invitrogen™ Supplier No. PA582424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AELKTAMSTP HVAEPAENEQ DEQDENGAEA SADLRADAMA KDRSEEERTT EAEKNERVQK HLKALTSELA NARDESKKTA NDMIHAENMR LGRDKYKTLR QIRQGNTKQR Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 98%.

Moesin, a member of the talin-4. 1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Moesin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MSN
Gene Accession No. O35763, P26038, P26041
Gene Alias C78546; epididymis luminal protein 70; HEL70; Membrane-organizing extension spike protein; MGC137877 protein; moesin; MSN
Gene Symbols MSN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MSN. Recombinant protein control fragment (Product #RP-90644).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17698, 4478, 81521
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.