Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOF / MYST1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993263
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3263 10 μL
50-199-3262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3263 Supplier Active Motif Supplier No. 61246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The MYST family of histone acetyltransferases, which includes MYST1, is named for the founding members MOZ (MYST3), yeast YBF2 and SAS2, and TIP60 (HTATIP). All members of this family contain a MYST region of about 240 amino acids with a canonical acetyl-CoA-binding site and a C2HC-type zinc finger motif. Most MYST proteins also have a chromodomain involved in protein-protein interactions and targeting transcriptional regulators to chromatin.

Specifications

Antigen MOF / MYST1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene KAT8
Gene Accession No. Q9H7Z6
Gene Alias 2010203C02Rik; 5830450F21Rik; D7Ertd629e; h FLJ14040; Histone acetyltransferase KAT8; histone acetyltransferase MYST1; hMOF; K (lysine) acetyltransferase 8; K(lysine) acetyltransferase 8; Kat8; Lysine acetyltransferase 8; Mof; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1; MYST histone acetyltransferase 1; MYST protein 1; MYST1; MYST-1; ortholog of Drosophila males absent on the first (MOF); PP7073; probable histone acetyltransferase MYST1; ZC2HC8
Gene Symbols KAT8
Host Species Rabbit
Immunogen This MOF / MYST1 antibody was raised against a peptide within the N-terminal region of human MOF / MYST1 (MYST histone acetyltransferase 1).
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84148
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.