Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOG Antibody (013), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289453MFV610
Description
MOG Monoclonal antibody specifically detects MOG in Human samples. It is validated for ELISASpecifications
MOG | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
013 | |
ELISA | |
MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human MOG (Accession#: NP_996532.2; Gly 30-Tyr 149). | |
0.1 mL | |
Cellular Markers | |
4340 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction