Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOG Monoclonal Antibody (C2), Invitrogen™

Catalog No. FEMA550880
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550880 20 μL
FEMA550879 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550880 Supplier Invitrogen™ Supplier No. MA550880
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The product of this gene is a membrane protein expressed on the oligodendrocyte cell surface and the outermost surface of myelin sheaths. Due to this localization, it is a primary target antigen involved in immune-mediated demyelination. This protein may be involved in completion and maintenance of the myelin sheath and in cell-cell communication. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOG
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene MOG
Gene Accession No. Q16653, Q63345
Gene Alias B230317G11Rik; BTN6; BTNL11; DAQB-92E24.2; dystonia 2, torsion (autosomal recessive); MGC26137; MOG; MOG alpha 6; MOG alpha-5; MOG alpha-6; MOG AluA; MOG AluB; MOG Ig AluB; MOG Ig-AluB; MOGIG2; MOGIG-2; myelin oligodendrocyte glycoprotein; myelin-oligodendrocyte glycoprotein; NRCLP7
Gene Symbols MOG
Host Species Mouse
Immunogen Recombinant Human MOG protein, Gly30-Tyr149 (Accession #Q16653).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100141421, 24558, 4340
Target Species Human, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.