Learn More
Invitrogen™ MOG Monoclonal Antibody (CL2858)

Description
Immunogen sequence: ALVGDEVELP CRISPGKNAT GMEVGWYRPP FSRVVHLYRN GKDQDGDQAP EYRGRTELLK DAIGEGKVTL RIRNVRFSDE GGFTCFFRDH SYQEEAAMEL KVEDPFYWVS PG Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%. Binds to an epitope located within the peptide sequence GMEVGWYRPP as determined by overlapping synthetic peptides.
Specifications
Specifications
| Antigen | MOG |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | CL2858 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MOG |
| Gene Accession No. | Q16653, Q61885, Q63345 |
| Gene Alias | B230317G11Rik; BTN6; BTNL11; DAQB-92E24.2; dystonia 2, torsion (autosomal recessive); MGC26137; MOG; MOG alpha 6; MOG alpha-5; MOG alpha-6; MOG AluA; MOG AluB; MOG Ig AluB; MOG Ig-AluB; MOGIG2; MOGIG-2; myelin oligodendrocyte glycoprotein; myelin-oligodendrocyte glycoprotein; NRCLP7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.