Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOGAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562101 Supplier Invitrogen™ Supplier No. PA562101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PIPVRQTLNP TQEQIEELHQ TYMEELRKLF EEHKGKYGIP EHETLV Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

Catalyzes the formation of diacylglycerol from 2-monoacylglycerol and fatty acyl-CoA. Probably not involved in absorption of dietary fat in the small intestine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOGAT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MOGAT1
Gene Accession No. Q96PD6
Gene Alias 0610030A14Rik; 1110064N14Rik; 2-acylglycerol O-acyltransferase 1; acyl CoA:monoacylglycerol acyltransferase 1; Acyl-CoA:monoacylglycerol acyltransferase 1; DC2; DGAT2L; Dgat2l1; Diacylglycerol acyltransferase 2-like protein 1; diacylglycerol O-acyltransferase 2 like 1; diacylglycerol O-acyltransferase 2-like 1; diacylglycerol O-acyltransferase candidate 2; hDC2; LOW QUALITY PROTEIN: 2-acylglycerol O-acyltransferase 1; mDC2; MGAT1; Mogat1; Monoacylglycerol O-acyltransferase 1; putative monoacylglycerol acyltransferase 1; WI1-2612I11.1
Gene Symbols MOGAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MOGAT1. Recombinant protein control fragment (Product #RP-100924).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.