Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MOGAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145135
Description
Immunogen sequence: GNFSVNYSDF KDLFPGFTSY LHVLPLWFWC PVFREYVMSV GLVSVSKKSV SYMVSKEGGG NIS Antigen sequence identity: Mouse 68%; Rat 73%.
Catalyzes the formation of diacylglycerol from 2-monoacylglycerol and fatty acyl-CoA. Probably not involved in absorption of dietary fat in the small intestine.
Specifications
MOGAT1 | |
Polyclonal | |
Unconjugated | |
MOGAT1 | |
0610030A14Rik; 1110064N14Rik; 2-acylglycerol O-acyltransferase 1; acyl CoA:monoacylglycerol acyltransferase 1; Acyl-CoA:monoacylglycerol acyltransferase 1; DC2; DGAT2L; Dgat2l1; Diacylglycerol acyltransferase 2-like protein 1; diacylglycerol O-acyltransferase 2 like 1; diacylglycerol O-acyltransferase 2-like 1; diacylglycerol O-acyltransferase candidate 2; hDC2; LOW QUALITY PROTEIN: 2-acylglycerol O-acyltransferase 1; mDC2; MGAT1; Mogat1; Monoacylglycerol O-acyltransferase 1; putative monoacylglycerol acyltransferase 1; WI1-2612I11.1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
116255 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PD6 | |
MOGAT1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 120-182. Recombinant protein control fragment (Product #RP-109844). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction