Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOGAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519372
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519372 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519372 Supplier Invitrogen™ Supplier No. PA519372
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Dietary fat absorption from the small intestine is facilitated by acyl-CoA:monoacylglycerol transferase (MOGAT; EC 2.3.1.22) and acyl-CoA:diacylglycerol acyltransferase (DGAT; see MIM 604900) activities. MOGAT catalyzes the joining of monoacylglycerol and fatty acyl-CoAs to form diacylglycerol (Yen and Farese, 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOGAT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MOGAT2
Gene Accession No. Q3SYC2
Gene Alias 2-acylglycerol O-acyltransferase 2; acyl CoA:monoacylglycerol acyltransferase 2; acyl-CoA:monoacylglycerol acyltransferase 2; DC5; Dgat2l5; DGAT2L5.; Diacylglycerol acyltransferase 2-like protein 5; diacylglycerol O-acyltransferase candidate 5; FLJ22644; hDC5; hMGAT2; Mgat1l; MGAT2; MGC119183; MGC119184; MGC119185; MOGAT2; Monoacylglycerol O-acyltransferase 1-like; Monoacylglycerol O-acyltransferase 2
Gene Symbols MOGAT2
Host Species Goat
Immunogen Synthetic peptide sequence (KESAAHILNRK) corresponding to the internal amino acids of MOGAT2 (aa 165-176).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80168
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.