Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOGAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583035 Supplier Invitrogen™ Supplier No. PA583035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPIFSFGEND LFDQIPNSSG SWLRYIQNRL QKIMGISLPL FHGRGVFQYS FGLIPYRRPI TTVVGKPIEV QKTLHPSEEE VNQLHQRYIK ELCNLFEAHK LKFNIPADQH LEF Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 81%.

Dietary fat absorption from the small intestine is facilitated by acyl-CoA:monoacylglycerol transferase (MOGAT; EC 2.3.1.22) and acyl-CoA:diacylglycerol acyltransferase (DGAT; see MIM 604900) activities. MOGAT catalyzes the joining of monoacylglycerol and fatty acyl-CoAs to form diacylglycerol (Yen and Farese, 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOGAT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MOGAT2
Gene Accession No. Q3SYC2
Gene Alias 2-acylglycerol O-acyltransferase 2; acyl CoA:monoacylglycerol acyltransferase 2; acyl-CoA:monoacylglycerol acyltransferase 2; DC5; Dgat2l5; DGAT2L5.; Diacylglycerol acyltransferase 2-like protein 5; diacylglycerol O-acyltransferase candidate 5; FLJ22644; hDC5; hMGAT2; Mgat1l; MGAT2; MGC119183; MGC119184; MGC119185; MOGAT2; Monoacylglycerol O-acyltransferase 1-like; Monoacylglycerol O-acyltransferase 2
Gene Symbols MOGAT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MOGAT2. Recombinant protein control fragment (Product #RP-90407).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.