Learn More
Eisco™ Moh's Hardness Kit, 12 Pieces
SCI_ED
Catalog No. S02916 Shop All Eisco Products
Change view
Click to view available options
Quantity:
12 Pieces
Catalog No. Quantity
S02916 12 Pieces
1 options
Catalog No. S02916 Supplier Eisco™ Supplier No. ESNGKIT0012
Add to Cart
Add to Cart
The Eisco Labs Moh's Hardness Kit includes 9 rock and Mineral specimens, 1 in. each, geology activity for exploring and identifying minerals using moh's hardness scale. Ideal for classrooms and geology enthusiasts, promoting hands-on learning and practical mineral study.
  • The Mohs hardness test is a simple and useful way to identify minerals and assess their relative hardness. This kit includes everything you need to explore using Mohs hardness test to identify minerals
  • The kit includes Nine different minerals (1 inch), including Soapstone, Gypsum, Calcite, Fluorite, Apatite, Microcline, Milky Quartz, Corundum, and Topaz, allowing you to test and explore various hardness levels
  • The included key card identifies each of the different minerals included in the kit. Two unglazed porcelain streak plates - one white, and one black - are also included for performing streak tests
  • Great for classrooms - Perfect for home schooling, a fun home science activity, and is a great addition to any classroom or geology laboratory. Useful for demonstrations in identifying and classifying rocks and minerals - perfect for group activities
  • Each specimen is selected and hand processed by an actual geologist, ensuring each sample exhibits defining features
  • Eisco Tech cut rock and mineral samples are sourced from a variety of locations around the world and are processed in Victor, NY
Specifications
Product Type Hardness Kit
Quantity 12 Pieces
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov