Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mohawk homeobox Antibody (OTI6B12), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272758R
Description
Mohawk homeobox Monoclonal specifically detects Mohawk homeobox in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Mohawk homeobox | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MKX | |
Full length recombinant protein of human Mohawk homeobox (NP_775847) produced in E.coli. | |
0.1 mL | |
283078.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI6B12 | |
Western Blot, Immunohistochemistry | |
C10orf48, homeobox protein Mohawk, IFRX, Iroquois family related homeodomain protein, iroquois homeobox protein-like 1, IRXL1chromosome 10 open reading frame 48, MGC39616, mohawk homeobox | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction