Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mohawk homeobox Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310948100UL
Description
Mohawk homeobox Polyclonal specifically detects Mohawk homeobox in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Mohawk homeobox | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry | |
C10orf48, homeobox protein Mohawk, IFRX, Iroquois family related homeodomain protein, iroquois homeobox protein-like 1, IRXL1chromosome 10 open reading frame 48, MGC39616, mohawk homeobox | |
The immunogen is a synthetic peptide directed towards the middle region of human Mohawk homeobox (NP_775847). Peptide sequence IKSENSVIKAGVRPESRASEDYVAPPKYKSSLLNRYLNDSLRHVMATNTT | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
283078 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction