Learn More
Eisco™ Mohs Hardness Kit
SCI_ED
Catalog No. S24660 Shop All Eisco Products
Change view
Click to view available options
Mohs Hardness Kit
Catalog No.
S24660 Mohs Hardness Kit
1 options
Catalog No. S24660 Supplier Eisco™ Supplier No. ESNG0001
For use in field, laboratory, or for personal use to determine hardness of unidentified samples. Mohs Hardness Kit contains nine specimens with distinct hardness and key.
  • Classroom tool for hands-on demonstration of mineral hardness
  • Numbered and stored in sturdy, sectioned container for easy access and safety
Specifications
Product Type Hardness Kit
For Use With (Application) Test hardness of unknown mineral sample up to a hardness of 9
Includes Talc, gypsum, calcite, fluorite, apatite, feldspar, quartz, topaz, and corundum
Packaging Sectioned container
Quantity 9 Specimens
Specimen Size Approx. 1 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov