Learn More
Invitrogen™ MOK Polyclonal Antibody

Description
Immunogen sequence: KQSLKQEEDR PKRRGPAYVM ELPKLKLSGV VRLSSYSSPT LQSVLGSGTN GRVPVLRPLK CIPASKKTDP QKDL Highest antigen sequence indentity to the following orthologs: Mouse - 65%, Rat - 65%.
Specifications
Specifications
| Antigen | MOK |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MOK |
| Gene Accession No. | Q9UQ07 |
| Gene Alias | MAPK/MAK/MRK overlapping kinase; MAPK/MAK/MRK/ overlapping kinase; MOK; MOK protein kinase; Rage; RAGE1; RAGE-1; renal cell carcinoma antigen; renal tumor antigen; renal tumor antigen 1; Serine/threonine kinase 30; soluble RAGE; STK30; T/STK 30 |
| Gene Symbols | MOK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.