Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOK Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734165 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4165 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4165 Supplier Proteintech Group Inc Supplier No. 239261AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.

Specifications

Antigen MOK
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MOK
Gene Accession No. Q9UQ07
Gene Alias MOK, MOK protein kinase, RAGE, RAGE 1, RAGE1, renal tumor antigen, Renal tumor antigen 1
Gene Symbols MOK
Host Species Rabbit
Immunogen MOK Fusion Protein Ag20849
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5891
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.