Learn More
Moldex™ 8000 Series Half-Mask Facepieces
SDP
Soft, durable facepiece confroms to the contours of the face without pressure points to provide long lasting comfort
Specifications
Certifications/Compliance NIOSH
Connection Type Snap-in gasket
Material Thermoplastic Elastomer
Product Type Half Mask Facepiece
View More Specs
Products 3
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
18-999-1029
SDP
View Documents
Moldex™
8001
Small N/A N/A N/A
18-999-1031
SDP
View Documents
Moldex™
8003
Large N/A N/A N/A
18-999-1030
SDP
View Documents
Moldex™
8002
Medium N/A N/A N/A
Description
Description
Soft, durable facepiece confroms to the contours of the face without pressure points to provide long lasting comfort
  • Extra wide surface seals easily and stays sealed even when talking
  • Low profile design provides a large field of vision
  • Facepiece has no gaskets or yoke; fewer parts to maintain and inventory
  • Each cartridge includes a new inhalation valve and features snap-in mount that eliminates mis-threading
Specifications
Specifications
NIOSH
Snap-in gasket
Thermoplastic Elastomer
Half Mask Facepiece
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.