Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ 9000 Series Full Face Respirator Facepiece
SDP

Catalog No. 19384317 Shop All Moldex Metric Inc. Products
Click to view available options
Format:
Small
Medium
Large
This item is not returnable. View return policy

Catalog No. 19-384-317

Supplier: Moldex™ 9003

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

At less than 13 ounces, the 9000 Series respirator is ultra-lightweight. Workers will appreciate the comfort and longer wearability and employers will benefit from increased user acceptance.

  • A convenient ratchet fastening strap system and the stand-away head harness make putting the 9000 mask on and taking it off quick and easy. The strap tabs are securely molded directly to the flange for rugged use.
  • The 9000 boasts an extra-wide viewing lens, with an exclusive over-molded flange design. This eliminates the traditional clamping frame found on most other face pieces.
  • The 9000 is completely free of metal parts, making it ideal for mines, refineries and other work environments where metal may be hazardous.
  • Advanced design eliminates unnecessary parts. Moldex™'s overmolded flange and harness tabs, plus integrated exhalation assembly, make cleaning and maintaining the 9000 Series respirator a breeze.

Compliance

Lens meets ANSI™ Z87+.

Specifications

Certifications/Compliance ANSI Z87+
Material Polycarbonate Lens
Product Type Respirator Facepiece
Format Large
For Use With (Application) Mines, refineries and other industries where metal parts can create an explosion hazard
Harness Style Stand away
Includes Tear-away lens protector
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.