Learn More
Moldex™ Comets™ Reusable Ear Plugs
SDP
Catalog No. 191500136 Shop All Moldex Metric Inc. Products
Click to view available options
Corded:
Corded
Uncorded
Comets™ reusable ear plugs feature a triple flange design that provides maximum comfort even when worn for long periods of time.
  • User-friendly finger pocket makes insertion easy
  • Extra soft material and an air-cushioned tip provide all-day comfort
  • Soft triple flange adjusts to most size ear canals to hold the plug securely in place
  • 100% PVC-Free™
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class BL
Size Universal
For Use With (Application) Utilities, Manufacturing, Production, Mining, Oil and Gas, Transportation, and Other Various Industries
High Visibility No
Packaging Type Poly bag
Pairs per Package 50 Pairs/Pk.
Plug Material PVC-free
Plug Color Green
Noise Reduction Rating (NRR) 25 dB
Corded Uncorded
Plug Shape Flanged
Disposable Reusable
Metal Detectable No
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.