Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ M3 Cap-Mounted Ear Muff
SDP

Catalog No. 19120504 Shop All Moldex Metric Inc. Products
Click to view available options
:
Cap-Mount
This item is not returnable. View return policy
This item is not returnable. View return policy

M3 Cap-Mounted Ear Muff slots easily into most hard hats.

  • SoftCoat™ covering helps muffle transmitted noise
  • Molded grips make fitting easy and quick
  • Exclusive iridescent color and curvy shape
  • Lightweight plastic arms
  • Extra soft foam

Specifications

Color Iridescent
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Equipment) Hard Hats
Headband Material Plastic
For Use With (Application) Construction, Energy and Utilities, General Manufacturing, Mining, Oil and Gas, Transportation
High Visibility No
Model M3 Series
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 24 dB
Product Type Ear Muffs
Dielectric No
Style Hard Hat
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.