Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Moldex™ 8000 Series Pre-Assembled Multi-Gas Vapor Smart™ Cartridge Respirators
Shop All Moldex Metric Inc. Products

Click to view available options
Size:
Small
Medium
Description
- 8000 Series half-mask pre-assembled with 8600 multi-gas/vapor smart cartridges
- Simplifies inventory by reducing need to stock multiple cartridge types
- Approved for nine common gas/vapors, replaces up to five gas/vapor cartridges
- Ideal for use against multiple hazards in same workplace
- For oil-free paint sprays and pesticides
- Soft, durable facepiece and lightweight design with extra-wide sealing
- Exhalation valve positioned for better communication
- Cartridges have a snap-in mount for easy replacement and no misthreading
- Living hinge allows head harness to be folded comfortably around neck when not in use
- Low profile, compact design fits well under welding helmets and with safety glasses
- Maintenance made simple with very few replacement parts
1 Includes two 19-003-346 N95 particulate prefilters, two 18-110 retainers, and two 18-102 organic vapor cartridges.
Specifications
Specifications
| Cartridge Type | Multi-Gas, Vapor Smart™ Cartridges |
| Certifications (NIOSH) | NIOSH 42 CFR Part 84, TC-23C-1223 |
| Color | Gray |
| Material | Thermoplastic Elastomer |
| Product Type | Pre-Assembled Half-Mask Respirator |
| Size | Small |
| For Use With (Application) | Degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing |
| Includes | 8000 Facepiece, 8600 Multi-Gas, Vapor Smart™ Cartridges |
| Model | 8601 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction