Moldex™ Pura-Fit™ Foam Ear Plugs

NRR: 33dB

Manufacturer: Moldex™ 6800

 View more versions of this product

Catalog No. 17-384

  Check Availability
Add to Cart

    Specifications

    Product Type Pura-Fit™ Earplug
    Shape Tapered
    Style Corded
    Certifications/Compliance ANSI S13.19-1974
    Size One size fits to all
    Material Polyurethane Foam
    Noise Reduction Range (NRR) 33dB
    Color Green
    For Use With (Application) Suitable for most industrial environments

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit