Learn More
Moldex™ Pura-Fit™ Disposable Ear Plugs
SDP
Catalog Number 17385
Click to view available options
Corded:
Corded
Uncorded
Packaging Type:
Paper pouch
Poly bag
Pura-Fit™ ear plugs are tapered and have a longer shape for easier insertion and grasping . Special foam is formulated for softness and comfort.
  • Extra-soft, low-pressure foam for comfort and fit
  • Tapered shape for easy insertion and removal
  • Non-irritating smooth surface
  • Longer length for easy fitting and removal
ANSI S3.1974 Yes
Cord Color Green
Cord Material Vinyl
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Energy, Utilities, Manufacturing, Healthcare, Mining, Oil and Gas, Transportation, and Other Various Industries
High Visibility Yes
Packaging Type Poly bag
Pairs per Package 100 Pairs/Pk.
Plug Material Foam
Plug Color Green
Noise Reduction Rating (NRR) 33 dB
Corded Corded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No