Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Molecular BioProducts™ PCR Tubes and Plates

Catalog No. 21402167
Change view
Click to view available options
Color:
Assorted
Natural
Orange
Purple
Closure Type:
Domed Cap
Volume (Metric):
0.2 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type Volume (Metric)
21-402-167 Assorted Domed Cap 0.2 mL
21-402-168 Orange Domed Cap 0.2 mL
21-402-110 Purple Domed Cap 0.2 mL
21-403-150 Natural - 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-402-167 Supplier Thermo Scientific™ Supplier No. 3414A

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Molded from 100% virgin polypropylene

Tubes feature thin walls for optimal heat transfer and caps that seal securely to minimize evaporation. Plates feature elevated well rims to ensure uniform sealing.

  • PurePak™ packaging reduces contamination from reaching into a bulk bag of tubes by dividing tubes into 10 separate PurePaks
  • Certified RNase-, DNase-, DNA, and Pyrogen-free
  • Clear, nonsterile

TRUSTED_SUSTAINABILITY

Specifications

Color Assorted
Volume (Metric) 0.2 mL
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Product Type Thin Wall PCR Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.