Learn More
Thermo Scientific™ Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Catalog No. 21403180
Change view
Click to view available options
Color:
Assorted
Blue
Green
Red
Yellow
Sterility:
Non-sterile
Sterile
Capacity (Metric):
0.6 mL
1.5 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Capacity (Metric)
21-403-180 Red Non-sterile 1.5 mL
21-403-163 Assorted Sterile 0.6 mL
21-403-178 Assorted Non-sterile 1.5 mL
21-403-181 Blue Sterile 1.5 mL
21-403-184 Green Non-sterile 1.5 mL
21-403-186 Yellow Non-sterile 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 21-403-180 Supplier Thermo Scientific™ Supplier No. 3444R
  • With flat cap
  • No frosting or graduations
  • Certified RNase/DNase, RNA/DNA, ATP and pyrogen free
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Flat
Certifications/Compliance RNase, DNase, RNA, DNA, ATP, pyrogen-free
Capacity (Metric) 1.5 mL
Color Red
Material Polypropylene
Includes Flat cap
Packaging Bag
Sterility Non-sterile
Graduated No
Closure Type Flat Cap