Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Amplex™ Red Sphingomyelinase Assay Kit

Catalog No. A12220
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12220 500 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12220 Supplier Invitrogen™ Supplier No. A12220
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Detects very low concentrations of sphingomyelinase

The Amplex™ Red Sphingomyelinase Assay Kit provides a sensitive, rapid, and simple fluorometric method for detecting very low concentrations of sphingomyelinase using a fluorescence microplate reader or fluorometer.

  • Detects sphingomyelinase activity levels as low as 80 μU/mL in a 200 μL assay volume
  • Format allows for multiple time point measurements
  • Designed for minimal autofluorescence interference

In this enzyme-coupled assay, sphingomyelinase activity is monitored indirectly using 10-acetyl-3,7-dihydroxyphenoxazine (Amplex™ Red reagent), a sensitive fluorogenic probe for hydrogen peroxide. Sphingomyelinase hydrolyses the sphingomyelin to yield ceramide and phosphorylcholine. Alkaline phosphatase is added, which hydrolyzes phosphorylcholine to form choline. The choline is then oxidized by choline oxidase to form betaine and hydrogen peroxide. In the presence of horseradish peroxidase, the hydrogen peroxide reacts with Amplex™ Red reagent in a 1:1 stoichiometric ratio to generate the highly fluorescent product resorufin.

Because resorufin has absorption and fluorescence emission maxima of approximately 571 nm and 585 nm, respectively, there is little interference from autofluorescence in most biological samples.

Use Amplex™ Red Assays for a Broad Range of Investigations

A wide variety of validated Amplex™ Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex™ UltraRed Reagent (Cat. No. A36006), a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex™ Red/UltraRed Stop Reagent (Cat. No. A33855). The Amplex™ Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours. Custom assay design and packaging are also available.

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Enzyme and Metabolic Activity
For Use With (Equipment) Fluorometer, Microplate Reader
Product Type Amplex Red Assay Kit
Dye Type Amplex Red
Format 96-well plate
Product Line Amplex
Quantity 500 Assays
Shipping Condition Wet Ice
Excitation Wavelength 571 nm
Emission Wavelength 585 nm
Show More Show Less
I'm using an Amplex Red kit, the reagent changes color to pink almost immediately in my own Krebs-Ringer buffer but not in HBSS. Why is this?

The components of Krebs-Ringer buffer (salts) should not cause oxidation of the Amplex reagent (which, in the presence of peroxidase and H2O2 oxidizes to resorufin, which is pink in color and fluorescent). Try water alone (the water used to make the Krebs-Ringer buffer). Since Hank's Buffered Saline Solution is typically purchased rather than made in the lab, it likely would not have the same contaminant. Another option is to degas the buffer prior to use to removed dissolved oxygen radicals.

Can Amplex Red Assays be performed using cell lysates?

This is not recommended. The presence of endogenous proteases can complicate the assay by degrading the horseradish peroxidase (HRP). Endogenous peroxidases and antioxidants can modify the H2O2 required for the reaction, competing with HRP (and catalase) for the substrate.

The Amplex Red Assays are best performed with either purified enzymes or extracted H2O2 in a defined buffer system, extracellular solutions or body fluids (media, serum, etc.) that do not exhibit high levels of endogenous protease or oxidase activity and do not contain antioxidants.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.