Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DiSC3(5) (3,3'-Dipropylthiadicarbocyanine Iodide)

Catalog No. D306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D306 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D306 Supplier Invitrogen™ Supplier No. D306
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potentiometric probe

DiSC3(5) is a carbocyanine with a short (C3) alkyl tail. This cationic dye accumulates on hyperpolarized membranes and is translocated into the lipid bilayer.

Cell Analysis, Cell Metabolism, Cell Structure, Cell Viability, Proliferation & Function, Membranes (General) & Lipids

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type DiSC3(5)
Quantity 100 mg
Shipping Condition Room Temperature
Sub Cellular Localization Cell Membranes & Lipids

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.