Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELF™ 97 Endogenous Phosphatase Detection Kit

Catalog No. E6601
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E6601 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E6601 Supplier Invitrogen™ Supplier No. E6601
  • $477.00
    $391.14 / Each of 1

    Save $85.86

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Can be used to detect phosphatase activity

Upon hydrolysis the ELF 97 phosphatase substrate in the ELF 97 Endogenous Phosphatase Kit produces a bright and photostable yellow-green fluorescent precipitate at the site of enzymatic activity. This fluorescent precipitate has several unique spectral characteristics, including an extremely large Stokes shift, 180nm that makes it easily distinguishable from endogenous fluorescence.

Cell Analysis, Cellular Imaging, Drug Discovery & Development, Enzyme & Protein Activity Assays, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Immunohistochemistry (IHC), Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation & Analysis, Target & Lead Identification & Validation

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow-Green
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Phosphatase Assay
For Use With (Equipment) Fluorescence Microscope
Product Type Phosphatase Assay
Dye Type ELF 97
Form Liquid
Product Line ELF
Quantity 1 kit
Shipping Condition Wet Ice
Substrate ELF™ 97 phosphate
Substrate Properties Chemical Substrate
Substrate Type Phosphatase Substrate
Target Enzyme Phosphatases
Emission Wavelength 345/530
Show More Show Less
Can the ELF 97 reagent be applied to live cells?

ELF 97 substrate is cell impermeant. It may be used on live cells that exhibit phosphatase activity on the surface of the cells, but not intracellular phosphatase activity in live cells. The alternative product for live-cell detection of phosphatase activity is the cell-permeable Alkaline Phosphatase Live Stain (Cat. No. A14353).

Can ELF 97 stained cell/tissue samples be multiplexed with antibody labeling?

No. The ELF 97 reagent does not covalently attach to any cellular components and may be washed away with any subsequent antibody labeling.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.