Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FluoZin™-3, Tetrapotassium Salt, cell impermeant

Catalog No. F24194
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F24194 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F24194 Supplier Invitrogen™ Supplier No. F24194
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zn2+-selective indicator with structure that resembles fluo-4

The FluoZin™-3 indicator is a Zn2+-selective indicator with a structure that resembles fluo-4. FluoZin™-3 exhibits high Zn2+-binding affinity that is unperturbed by Ca2+ concentrations up to at least 1 μM. In addition, FluoZin™-3 exhibits a >50-fold increase in fluorescence in response to saturating levels of Zn2+. Although imaging applications of FluoZin™-3 are predominant, cell-based microplate assays and flow cytometry protocols have also been developed. FluoZin™-3 indicator is available in a cell-permeant and a cell-impermeant form.

Fluorescent Zinc Indicators Specifications:

  • Label (Ex/Em): FluoZin™-3 (∼494/516 nm)
  • Kd (Zn2+) (in buffer): ∼15 nM
  • Lyophilized product may be dissolved in distilled water or aqueous buffer for use
  • Product is typically loaded into cellsusing microinjection or diffusion from patch pipette

Find Fluorescent Indicators for Zinc and Other Metal Ions

We offer a number of fluorescent indicators to determine polyvalent cation concentrations inside cells, to follow metal ion transport through ion channels, to make measurements in environmental samples, and more. Review Fluorescent Indicators for Zn2+ and Other Metal Ions—Section 19.7 in the Molecular Probes™ Handbook for more information on these products.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Zinc Indicator
For Use With (Equipment) Fluorescence Microscope
Product Type Zinc Indicator
Dye Type FluoZin-3
Product Line FluoZin
Quantity 500 μg
Shipping Condition Room Temperature
Excitation Wavelength 494 nm
Emission Wavelength 516 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.