Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUN™ 1 Cell Stain

Catalog No. F7030
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F7030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F7030 Supplier Invitrogen™ Supplier No. F7030
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Two-color fluorescent viability probe for yeast and fungi.

FUN™ 1 is a unique two-color fluorescent viability probe for yeast and fungi. The FUN™ 1 stain passively diffuses into a variety of cell types and initially stains the cytoplasm with a diffusely distributed green fluorescence. However, in several common species of yeast and fungi, subsequent processing of the dye by live cells results in the formation of distinct vacuolar structures with compact form that exhibit a striking red fluorescence, accompanied by a reduction in the green cytoplasmic fluorescence. Formation of the intravacuolar structures requires both plasma membrane integrity and metabolic capability. Dead cells fluoresce bright yellow-green, with no discernable red structures.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Fungal Cells, Yeast Cells
Concentration 10 mM
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Yeast/Fungi Stain
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Stain
Dye Type FUN 1
Excitation Wavelength Range 480 to 495 nm
Format Tubes, Slides
Product Line FUN
Quantity 100 μL
Shipping Condition Room Temperature
Emission Wavelength Range 525 to 540 nm, 615 to 630 nm
Excitation Wavelength 485 nm
Emission Wavelength 530 nm, 620 nm
Show More Show Less
Can FUN 1 stained cells be examined by flow cytometry?

Yes. Use a 488 nm laser line and standard FITC and PE channels for two-color detection of green (dead/metabolically inactive cells) and red (live, metabolically active cells) emission.

Live, metabolically active fungi transport FUN 1 into vacuoles to give a red-shifted fluorescence versus green/yellow fluorescence in the nucleus and cytoplasm of dead or metabolically-inactive cells. Is this a reliable indicator of fungal viability?

No. FUN 1 accumulates into vacuoles by an unknown transport pathway, but any mutants/ recombinant cells or experimental treatments that result in a deficiency or block in vesicle-mediate transport into vacuoles may result in cells that do not have red vacuoles, even though the cells are live and metabolically active. For more information see J Microbiol Methods 78:208 (2009).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.