Learn More
Invitrogen™ Fura-4F, AM, cell permeant - Special Packaging
Description
- Acetoxymethyl (AM) ester derivative
- Useful for noninvasive intracellular loading
- Ex/Ems are identical to that of fura-2, and more importantly the Kd (Ca2+)μM for each are very similar: 0.77 for fura-4F AM
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Cell-Based Second Messenger Assays, Drug Discovery and Development, G-Protein Coupled Receptor Biology, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell Viability and Proliferation |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Dye |
| Dye Type | Fluorescent Dye-Based |
| Quantity | 10 x 50 μg |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).
After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.