Learn More
Invitrogen™ Gelatin From Pig Skin, Fluorescein Conjugate
Description
Molecular Probes fluorescent conjugates of gelatin are designed for researchers studying not only collagen-binding proteins and collagen metabolism, but also for the study of gelatinases and collagenases (metalloproteins that digest collagen and gelatin).
- Label (Ex/Em): Fluorescein (approx. 499/519nm)
- Degree of labeling is listed on the product label (based on molecular weight of 100,000)
Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Quantity | 5 mg |
| Conjugate | Fluorescein |
| Species | Pig |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Classic Dyes |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Gelatin |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.