Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Staphylococcus aureus (Wood strain without protein A) BioParticles™, Alexa Fluor™ 594 conjugate

Catalog No. S23372
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S23372 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23372 Supplier Invitrogen™ Supplier No. S23372
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescently labeled, heat- or chemically killed bacteria and yeast in a variety of sizes, shapes, and natural antigenicities

Fluorescent products have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry

Unlike the fluorescence of fluorescein-labeled BioParticles conjugates, which is partially quenched in acidic environments, the fluorescence of the Alexa Fluor BODIPY™ FL, tetramethylrhodamine and Texas Red™ dye conjugates is uniformly intense over the pH range from 3 to 10.

  • Label (Ex/Em): Alexa Fluor 594 (approx. 590/617nm)
  • Particle: S. aureus (Wood strain, without protein A)
  • Provided as lyophilized powders
  • approximately 3 x 108 S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid
  • Conjugates can be reconstituted in buffer of choice for use in phagocytosis assays
  • Conjugates that are bound to the surface of the cell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers
  • Fluorescent BioParticles conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants
  • Small particles may also be useful references for light scattering studies because their sizes and shapes differ in characteristic ways

Cell Analysis, Cell Structure, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Receptors

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Bioparticle Conjugate
Dye Type Alexa Fluor™ 594
Form Lyophilized Powder
pH 3 to 10
Product Line Alexa Fluor, BioParticles
Quantity 2 mg
Shipping Condition Room Temperature
Species S. aureus
Show More Show Less
Are the Invitrogen BioParticles products sterile?

While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.

What is the type of bond that attaches the dyes to the BioParticles probes?

We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.

What can the BioParticles product line be used for?

Fluorescent Bioparticles have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry. We offer E.Coli, S. aureus, and zymosan BioParticles conjugates covalently labeled with a variety of different fluorophores.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.