Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIVE BacLight™ Bacterial Gram Stain Kit, for microscopy & quantitative assays

Catalog No. L7005
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L7005 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L7005 Supplier Invitrogen™ Supplier No. L7005
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

One step assay

The LIVE BacLight™ Bacterial Gram Stain Kit provides a novel one-step fluorescence assay for the determination of gram sign in living bacteria. When gram negative and gram-positive bacterial are simultaneously stained with the membrane-permeant SYTO 9 dye and hexidium iodide, gram-negative bacteria fluoresce green and gram-positive bacteria fluoresce red.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green, Red
Content And Storage Store in freezer at -5°C to -30°C and protect from light.
Description LIVE BacLight™ Bacterial Gram Stain Kit, for microscopy and quantitative assays
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Fluorescence Microscope, Fluorometer, Flow Cytometer, Microplate Reader
Product Type Bacterial Stain Kit
Dye Type Other Label(s) or Dye(s)
Format Tube(s), 96-well plate, Slide(s)
Product Line BacLight
Quantity 1 kit
Shipping Condition Room Temperature
Excitation Wavelength 470 nm
Emission Wavelength 480 to 700
Show More Show Less
What bacterial parameters can I look at by flow cytometry?

You can stain bacteria with a general stain such as BacLight Green Bacterial Stain (Cat. No. B35000) or BacLight Red Bacterial Stain (Cat. No. B35001). You can look at gram character (Cat. No. L7005), cell viability (Cat. Nos. L7007, L7012, and L13152), cell count (Cat. Nos. L34856 and B7277), and cell vitality. Cell vitality can be measured by membrane potential (Cat. No. B34950) or by metabolism (Cat. Nos. B34954 and B34956).

What is the excitation/emission maxima for SYTO 9 stain and hexidium iodide?

The excitation/emission maxima for these dyes are 480/500 nm for SYTO 9 stain and 480/625 nm for hexidium iodide.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.