Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LysoSensor™ Yellow/Blue dextran, 10,000 MW, Anionic, Fixable

Catalog No. L22460
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L22460 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L22460 Supplier Invitrogen™ Supplier No. L22460
  • $550.00
    $451.00 / Each of 1

    Save $99.00

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Ratiometric probe

LysoSensor Yellow/Blue dextran can be used to measure the pH of acidic organelles, such as lysosomes. This dextran conjugate is taken up by the cells through endocytosis. The LysoSensor dye produces blue fluorescence in neutral environments, but changes to yellow fluorescence in more acidic environments. The pKa of LysoSensor Yellow/Blue dextran is ∽3.9.

Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Ionic Homeostasis & Signaling, Lysosomes, Organelle Tracing, pH Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow, Blue
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Dextran
Product Line LysoSensor
Quantity 5 mg
Shipping Condition Room Temperature
Sub Cellular Localization Lysosomes
Emission Wavelength 335/452
Can I use a plate reader to measure the pH of acidic organelles, such as lysosomes, with LysoSensor ratiometric probes?

Using a plate reader for measuring the pH of acidic organelles, such as lysosomes, with LysoSensor ratiometric probes, provides weak pH measurements, as discrete regions of interest cannot be selected for analysis. Therefore, there is no way of excluding extracellular fluorescence and little to no spatial or morphological information can be obtained. Instead, a readout of whole-cell fluorescence is provided, resulting in the average intraorganellar pH of all labeled organelles for the analyzed cell population. The is also the risk that a signal may be dominated by a few extremely bright, but essentially irrelevant cells, such as dead cells, or dye-labeled debris particles.

For more information about measuring luminal pH in organelles, please see the reference linked below:

Live-cell Microscopy and Fluorescence-based Measurement of Luminal pH in Intracellular Organelles

What reagents would you recommend for measuring the pH in acidic organelles, such as lysosomes?

For measuring the pH in acidic organelles, such as lysosomes, we recommend our LysoSensor ratio metric probes. Please see the links to the product page and descriptions below:

LysoSensor Yellow/Blue dextran, 10,000 MW, Anionic, Fixable (Cat. No. L22460):
LysoSensor Yellow/Blue dextran is a ratiometric probe that can be used to measure the pH of acidic organelles, such as lysosomes. This dextran conjugate is taken up by the cells through endocytosis. The LysoSensor dye produces blue fluorescence in neutral environments, but changes to yellow fluorescence in more acidic environments. The pKa of LysoSensor Yellow/Blue dextran is ~3.9.

LysoSensor Yellow/Blue DND-160 - Special Packaging (Cat. No. L7545):
LysoSensor Yellow/Blue DND-160 is a ratiometric probe that can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye produces blue fluorescence in neutral environments, but changes to yellow fluorescence in more acidic environments. The pKa of LysoSensor Yellow/Blue DND-160 is ~4.2.

LysoSensor Blue DND-167 - Special Packaging (Cat. No. L7533):
LysoSensor Blue DND-167 can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye becomes more fluorescent in acidic environments. The pKa of LysoSensor Blue DND-167 is ~5.1.

LysoSensor Green DND-189 - Special Packaging (Cat. No. L7535):
LysoSensor Green DND-189 can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye becomes more fluorescent in acidic environments. The pKa of LysoSensor Green DND-189 is ~5.2.

Could you provide me a protocol or reference for measuring the pH in acidic organelles, such as lysosomes?

Please find a link to a reference for measuring luminal pH in intracellular organelles below:

Live-cell Microscopy and Fluorescence-based Measurement of Luminal pH in Intracellular Organelles

What is the difference between LysoSensor Yellow/Blue dextran, 10,000 MW, Anionic, Fixable (Cat. No. L22460) and LysoSensor Yellow/Blue DND-160 - Special Packaging (Cat. No. L7545)?

LysoSensor Yellow/Blue dextran, 10,000 MW, Anionic, Fixable (Cat. No. L22460) has an emission of 335/452, is fixable and allows loading of cells by endocytosis. The pKa of LysoSensor Yellow/Blue dextran is ~3.9, which is slightly lower than LysoSensor Yellow/Blue DND-160 - Special Packaging (Cat. No. L7545).

LysoSensor Yellow/Blue DND-160 - Special Packaging (Cat. No. L7545) has an emission of 329/440 and is unique as it shows both dual-excitation and dual-emission spectral peaks that are pH-dependent. Nevertheless, this LysoSensor only exhibits the pH-dependent dual-emission spectra in living cells. Lysosensor Yellow/Blue DND-160 has predominantly yellow fluorescence, and in less acidic organelles is has blue fluorescence. Dual-emission measurements may permit ratio imaging of the pH in acidic organelles such as lysosome.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.